Methyl 2-Acetamido-3-(1,2,4-triazol-1-yl)propanoate

Lieferant: Apollo Scientific

OR470696-1G
APOSOR470696-1GEA 916 EUR
APOSOR470696-1G
Methyl 2-Acetamido-3-(1,2,4-triazol-1-yl)propanoate
Methyl 2-Acetamido-3-(1,2,4-triazol-1-yl)propanoate
Formel: C₈H₁₂N₄O₃
Molecular Weight: 212,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21236282
CAS-Nummer: 1487253-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR