Methyl 2-Acetamido-3-(1-pyrazolyl)propanoate
Lieferant: Apollo Scientific
OR470697-1G
APOSOR470697-1GEA
916
EUR
APOSOR470697-1G
Methyl 2-Acetamido-3-(1-pyrazolyl)propanoate
Methyl 2-Acetamido-3-(1-pyrazolyl)propanoate
Formel:
C₉H₁₃N₃O₃ Molecular Weight: 211,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21236281 CAS-Nummer: 170305-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...