2-Amino-3-(1-pyrazolyl)propanoic acid

Lieferant: Apollo Scientific

OR470698-1G
APOSOR470698-1GEA 804 EUR
APOSOR470698-1G
2-Amino-3-(1-pyrazolyl)propanoic acid
2-Amino-3-(1-pyrazolyl)propanoic acid
Formel: C₆H₉N₃O₂
Molecular Weight: 155,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832221
CAS-Nummer: 28024-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR