4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester
Lieferant: Apollo Scientific
OR470702-5G
APOSOR470702-5GEA
360
EUR
APOSOR470702-5G
4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester
4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester
Formel:
C₁₆H₂₁BN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18735902 CAS-Nummer: 1351478-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...