2-(Allyloxy)-4-methoxybenzyl chloride

Lieferant: Apollo Scientific

OR470703-1G
APOSOR470703-1GEA 916 EUR
APOSOR470703-1G
2-(Allyloxy)-4-methoxybenzyl chloride
2-(Allyloxy)-4-methoxybenzyl chloride
Formel: C₁₁H₁₃ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD26937559
CAS-Nummer: 1823868-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR