2-(Allyloxy)-4-nitrobenzyl chloride

Lieferant: Apollo Scientific

OR470704-1G
APOSOR470704-1GEA 916 EUR
APOSOR470704-1G
2-(Allyloxy)-4-nitrobenzyl chloride
2-(Allyloxy)-4-nitrobenzyl chloride
Formel: C₁₀H₁₀ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD26937558
CAS-Nummer: 1823885-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR