Diethyl 2-(4-Methoxybenzyl)malonate

Lieferant: Apollo Scientific

OR470705-5G
APOSOR470705-5GEA 601 EUR
APOSOR470705-5G
Diethyl 2-(4-Methoxybenzyl)malonate
Diethyl 2-(4-Methoxybenzyl)malonate
Formel: C₁₅H₂₀O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00157074
CAS-Nummer: 6335-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR