3-Cyano-5-(methoxycarbonyl)benzoic acid

Lieferant: Apollo Scientific

OR470706-1G
APOSOR470706-1GEA 1120 EUR
APOSOR470706-1G
3-Cyano-5-(methoxycarbonyl)benzoic acid
3-Cyano-5-(methoxycarbonyl)benzoic acid
Formel: C₁₀H₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08458980
CAS-Nummer: 126739-90-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR