3,3-Dimethyl-1-oxoisoindoline-5-carbonitrile

Lieferant: Apollo Scientific

OR470708-1G
APOSOR470708-1GEA 1120 EUR
APOSOR470708-1G
3,3-Dimethyl-1-oxoisoindoline-5-carbonitrile
3,3-Dimethyl-1-oxoisoindoline-5-carbonitrile
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23703554
CAS-Nummer: 1440519-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR