Methyl 3-(4-Hydroxy-2-methylphenyl)propanoate

Lieferant: Apollo Scientific

OR470710-1G
APOSOR470710-1GEA 916 EUR
APOSOR470710-1G
Methyl 3-(4-Hydroxy-2-methylphenyl)propanoate
Methyl 3-(4-Hydroxy-2-methylphenyl)propanoate
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11616255
CAS-Nummer: 105731-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR