2-Chloro-4-phenylquinazoline

Lieferant: Apollo Scientific

OR470712-5G
APOSOR470712-5GEA 0 EUR
APOSOR470712-5G
2-Chloro-4-phenylquinazoline
2-Chloro-4-phenylquinazoline
Formel: C₁₄H₉ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01152724
CAS-Nummer: 29874-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR