3-Chloro-5-(methoxycarbonyl)benzoic acid
Lieferant: Apollo Scientific
OR470713-1G
APOSOR470713-1GEA
713
EUR
APOSOR470713-1G
3-Chloro-5-(methoxycarbonyl)benzoic acid
3-Chloro-5-(methoxycarbonyl)benzoic acid
Formel:
C₉H₇ClO₄ Molecular Weight: 214,602 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458982 CAS-Nummer: 153203-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...