Methyl Morpholine-4-carboxylate

Lieferant: Apollo Scientific

OR470715-5G
APOSOR470715-5GEA 916 EUR
APOSOR470715-5G
Methyl Morpholine-4-carboxylate
Methyl Morpholine-4-carboxylate
Formel: C₆H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00595228
CAS-Nummer: 6906-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR