(2,4-Dichlorobenzyl)hydrazine dihydrochloride

Lieferant: Apollo Scientific

OR470716-5G
APOSOR470716-5GEA 1210 EUR
APOSOR470716-5G
(2,4-Dichlorobenzyl)hydrazine dihydrochloride
(2,4-Dichlorobenzyl)hydrazine dihydrochloride
Formel: C₇H₈Cl₂N₂·2ClH
Molecular Weight: 263,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26937552
CAS-Nummer: 1469998-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR