2'-(Allyloxy)acetophenone

Lieferant: Apollo Scientific

OR470717-5G
APOSOR470717-5GEA 601 EUR
APOSOR470717-5G
2'-(Allyloxy)acetophenone
2'-(Allyloxy)acetophenone
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026218
CAS-Nummer: 53327-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR