2-(2-Methyl-1-piperazinyl)acetic acid

Lieferant: Apollo Scientific

OR470718-1G
APOSOR470718-1GEA 916 EUR
APOSOR470718-1G
2-(2-Methyl-1-piperazinyl)acetic acid
2-(2-Methyl-1-piperazinyl)acetic acid
Formel: C₇H₁₄N₂O₂
Molecular Weight: 158,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698344
CAS-Nummer: 1060813-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR