Methyl 2-Methyl-3-oxo-3-phenylpropanoate

Lieferant: Apollo Scientific

OR470721-5G
APOSOR470721-5GEA 713 EUR
APOSOR470721-5G
Methyl 2-Methyl-3-oxo-3-phenylpropanoate
Methyl 2-Methyl-3-oxo-3-phenylpropanoate
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00956709
CAS-Nummer: 29540-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR