1,7-Dimethyl-1,4-diazepane
Lieferant: Apollo Scientific
OR470722-1G
APOSOR470722-1GEA
804
EUR
APOSOR470722-1G
1,7-Dimethyl-1,4-diazepane
1,7-Dimethyl-1,4-diazepane
Formel:
C₇H₁₆N₂ Molecular Weight: 128,215 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19221307 CAS-Nummer: 67744-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...