Methyl (1R,2S,3S,4S,6R)-5-oxotricyclo[2.2.1.02,6]heptane-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: Tricyclo[2.2.1.02,6]heptane-3-carboxylic acid, 5-oxo-, methyl ester, stereoisomer

OR470727-1G
APOSOR470727-1GEA 804 EUR
APOSOR470727-1G
Methyl (1R,2S,3S,4S,6R)-5-oxotricyclo[2.2.1.02,6]heptane-3-carboxylate
Methyl (1R,2S,3S,4S,6R)-5-oxotricyclo[2.2.1.02,6]heptane-3-carboxylate
Formel: C₉H₁₀O₃
Molecular Weight: 166,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28096495
CAS-Nummer: 1932427-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR