(1R,2S,3S,4S,6R)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylic acid

Lieferant: Apollo Scientific

OR470728-1G
APOSOR470728-1GEA 713 EUR
APOSOR470728-1G
(1R,2S,3S,4S,6R)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylic acid
(1R,2S,3S,4S,6R)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylic acid
Formel: C₈H₈O₃
Molecular Weight: 152,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08310685
CAS-Nummer: 71155-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR