Methyl (1S,2R,3R,4R,6S)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylate

Lieferant: Apollo Scientific

OR470729-1G
APOSOR470729-1GEA 713 EUR
APOSOR470729-1G
Methyl (1S,2R,3R,4R,6S)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylate
Methyl (1S,2R,3R,4R,6S)-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylate
Formel: C₉H₁₀O₃
Molecular Weight: 166,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28023496
CAS-Nummer: 176019-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR