2-(Allyloxy)-4-methoxybenzyl Bromide

Lieferant: Apollo Scientific

OR470730-1G
APOSOR470730-1GEA 916 EUR
APOSOR470730-1G
2-(Allyloxy)-4-methoxybenzyl Bromide
2-(Allyloxy)-4-methoxybenzyl Bromide
Formel: C₁₁H₁₃BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD26937561
CAS-Nummer: 1823942-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR