2-(Allyloxy)-4-nitrobenzyl Bromide

Lieferant: Apollo Scientific

OR470731-1G
APOSOR470731-1GEA 1120 EUR
APOSOR470731-1G
2-(Allyloxy)-4-nitrobenzyl Bromide
2-(Allyloxy)-4-nitrobenzyl Bromide
Formel: C₁₀H₁₀BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD26937560
CAS-Nummer: 1823359-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR