Methyl-5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 5-bromoorotate, Methyl 5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate

OR470732-1G
APOSOR470732-1GEA 1120 EUR
APOSOR470732-1G
Methyl-5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Methyl-5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Formel: C₆H₅BrN₂O₄
Molecular Weight: 249,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07023034
CAS-Nummer: 30825-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR