1,4-Dimethylimidazole-5-carboxylic acid

Lieferant: Apollo Scientific

OR470733-1G
APOSOR470733-1GEA 601 EUR
APOSOR470733-1G
1,4-Dimethylimidazole-5-carboxylic acid
1,4-Dimethylimidazole-5-carboxylic acid
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063163
CAS-Nummer: 78449-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR