3-Formyl-4-methylbenzoic acid

Lieferant: Apollo Scientific

OR470735-5G
APOSOR470735-5GEA 916 EUR
APOSOR470735-5G
3-Formyl-4-methylbenzoic acid
3-Formyl-4-methylbenzoic acid
Formel: C₉H₈O₃
Molecular Weight: 164.158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20660432
CAS-Nummer: 69526-89-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR