2-Cyano-3-(4-hydroxyphenyl)propanoic acid

Lieferant: Apollo Scientific

OR470737-5G OR470737-1G
APOSOR470737-5GEA 1330 EUR
APOSOR470737-5G APOSOR470737-1G
2-Cyano-3-(4-hydroxyphenyl)propanoic acid
2-Cyano-3-(4-hydroxyphenyl)propanoic acid
Formel: C₁₀H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11593027
CAS-Nummer: 90924-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR