2-Cyano-3-(4-hydroxy-3-methoxyphenyl)propanoic acid
Lieferant: Apollo Scientific
OR470738-5G
APOSOR470738-5GEA
1330
EUR
APOSOR470738-5G
2-Cyano-3-(4-hydroxy-3-methoxyphenyl)propanoic acid
2-Cyano-3-(4-hydroxy-3-methoxyphenyl)propanoic acid
Formel:
C₁₁H₁₁NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11593025 CAS-Nummer: 1082386-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...