(Isopropylamino)(oxo)acetic acid

Lieferant: Apollo Scientific

Synonyme: 2-(Isopropylamino)-2-oxoacetic acid

OR470739-5G
APOSOR470739-5GEA 567 EUR
APOSOR470739-5G
(Isopropylamino)(oxo)acetic acid
(Isopropylamino)(oxo)acetic acid
Formel: C₅H₉NO₃
Molecular Weight: 131,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06740186
CAS-Nummer: 29262-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR