3-Chloro-5-(hydroxymethyl)benzoic acid

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-5-(hydroxymethyl)benzoic Acid

OR470740-1G
APOSOR470740-1GEA 478 EUR
APOSOR470740-1G
3-Chloro-5-(hydroxymethyl)benzoic acid
3-Chloro-5-(hydroxymethyl)benzoic acid
Formel: C₈H₇ClO₃
Molecular Weight: 186,592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20658596
CAS-Nummer: 153203-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR