4-Amino-2-(Boc-aminomethyl)pyridine
Lieferant: Apollo Scientific
OR470742-1G
APOSOR470742-1GEA
1120
EUR
APOSOR470742-1G
4-Amino-2-(Boc-aminomethyl)pyridine
4-Amino-2-(Boc-aminomethyl)pyridine
Formel:
C₁₁H₁₇N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07374899 CAS-Nummer: 886371-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...