Benzofuran-7-carbonitrile
Lieferant: Apollo Scientific
OR470745-1G
APOSOR470745-1GEA
713
EUR
APOSOR470745-1G
Benzofuran-7-carbonitrile
Benzofuran-7-carbonitrile
Formel:
C₉H₅NO Molecular Weight: 143,142 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15143950 CAS-Nummer: 52951-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...