5-Chloro-N,N-diethyl-1-pentanamine hydrochloride
Lieferant: Apollo Scientific
OR470747-1G
APOSOR470747-1GEA
1120
EUR
APOSOR470747-1G
5-Chloro-N,N-diethyl-1-pentanamine hydrochloride
5-Chloro-N,N-diethyl-1-pentanamine hydrochloride
Formel:
C₉H₂₀ClN·ClH Molecular Weight: 214,176 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27500642 CAS-Nummer: 90796-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...