4-Chloro-N,N-diethyl-1-butanamine hydrochloride
Lieferant: Apollo Scientific
OR470749-1G
APOSOR470749-1GEA
601
EUR
APOSOR470749-1G
4-Chloro-N,N-diethyl-1-butanamine hydrochloride
4-Chloro-N,N-diethyl-1-butanamine hydrochloride
Formel:
C₈H₁₈ClN·ClH Molecular Weight: 200,149 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27500641 CAS-Nummer: 108130-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...