Methyl 2,5-Diaminobenzoate
Lieferant: Apollo Scientific
OR470750-1G
APOSOR470750-1GEA
503
EUR
APOSOR470750-1G
Methyl 2,5-Diaminobenzoate
Methyl 2,5-Diaminobenzoate
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12913485 CAS-Nummer: 49592-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...