4-(Azetidine-1-carbonyl)phenylboronic acid

Lieferant: Apollo Scientific

OR470751-1G
APOSOR470751-1GEA 1120 EUR
APOSOR470751-1G
4-(Azetidine-1-carbonyl)phenylboronic acid
4-(Azetidine-1-carbonyl)phenylboronic acid
Formel: C₁₀H₁₂BNO₃
Molecular Weight: 205,018 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22200200
CAS-Nummer: 1025664-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR