2-(2-Thienyl)imidazo[1,2-a]pyridine-7-carboxylic acid
Lieferant: Apollo Scientific
OR470752-1G
APOSOR470752-1GEA
1210
EUR
APOSOR470752-1G
2-(2-Thienyl)imidazo[1,2-a]pyridine-7-carboxylic acid
2-(2-Thienyl)imidazo[1,2-a]pyridine-7-carboxylic acid
Formel:
C₁₂H₈N₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD08164315 CAS-Nummer: 928003-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...