2-Phenylimidazo[1,2-a]pyridine-7-carboxylic acid
Lieferant: Apollo Scientific
OR470753-1G
APOSOR470753-1GEA
997
EUR
APOSOR470753-1G
2-Phenylimidazo[1,2-a]pyridine-7-carboxylic acid
2-Phenylimidazo[1,2-a]pyridine-7-carboxylic acid
Formel:
C₁₄H₁₀N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08164306 CAS-Nummer: 959-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...