Methyl 1,4,5,6-Tetrahydropyridine-3-carboxylate

Lieferant: Apollo Scientific

OR470754-1G
APOSOR470754-1GEA 1940 EUR
APOSOR470754-1G
Methyl 1,4,5,6-Tetrahydropyridine-3-carboxylate
Methyl 1,4,5,6-Tetrahydropyridine-3-carboxylate
Formel: C₇H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05669331
CAS-Nummer: 14997-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR