3,5-Dibromo-4-chlorophenol

Lieferant: Apollo Scientific

OR470759-5G
APOSOR470759-5GEA 1120 EUR
APOSOR470759-5G
3,5-Dibromo-4-chlorophenol
3,5-Dibromo-4-chlorophenol
Formel: C₆H₃Br₂ClO
Molecular Weight: 286,348 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18914596
CAS-Nummer: 74104-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR