4-(Aminomethyl)-2,2-dimethyl-1,3-benzodioxole

Lieferant: Apollo Scientific

Synonyme: (2,2-Dimethylbenzo[d][1,3]dioxol-4-yl)methanamine

OR470761-1G
APOSOR470761-1GEA 916 EUR
APOSOR470761-1G
4-(Aminomethyl)-2,2-dimethyl-1,3-benzodioxole
4-(Aminomethyl)-2,2-dimethyl-1,3-benzodioxole
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27500636
CAS-Nummer: 1823947-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR