2-Amino-5-(chloromethyl)pyrimidine hydrochloride

Lieferant: Apollo Scientific

OR470763-1G
APOSOR470763-1GEA 1120 EUR
APOSOR470763-1G
2-Amino-5-(chloromethyl)pyrimidine hydrochloride
2-Amino-5-(chloromethyl)pyrimidine hydrochloride
Formel: C₅H₆ClN₃·ClH
Molecular Weight: 180,035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09056752
CAS-Nummer: 120747-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR