4-Methoxy-6-methylsalicylaldehyde

Lieferant: Apollo Scientific

OR470764-1G
APOSOR470764-1GEA 260 EUR
APOSOR470764-1G
4-Methoxy-6-methylsalicylaldehyde
4-Methoxy-6-methylsalicylaldehyde
Formel: C₉H₁₀O₃
Molecular Weight: 166,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496263
CAS-Nummer: 34883-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR