2-(Hydroxymethyl)-2-phenyl-1,3-propanediol

Lieferant: Apollo Scientific

OR470765-5G
APOSOR470765-5GEA 855 EUR
APOSOR470765-5G
2-(Hydroxymethyl)-2-phenyl-1,3-propanediol
2-(Hydroxymethyl)-2-phenyl-1,3-propanediol
Formel: C₁₀H₁₄O₃
Molecular Weight: 182,216 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18642376
CAS-Nummer: 4704-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR