1-(Chloromethyl)-4-methylcyclohexane

Lieferant: Apollo Scientific

OR470766-1G
APOSOR470766-1GEA 916 EUR
APOSOR470766-1G
1-(Chloromethyl)-4-methylcyclohexane
1-(Chloromethyl)-4-methylcyclohexane
Formel: C₈H₁₅Cl
Lagertemperatur: Raumtemperatur
MDL: MFCD16657516
CAS-Nummer: 1073-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR