1-(Bromomethyl)-4-(tert-butyl)cyclohexane

Lieferant: Apollo Scientific

OR470767-1G
APOSOR470767-1GEA 1120 EUR
APOSOR470767-1G
1-(Bromomethyl)-4-(tert-butyl)cyclohexane
1-(Bromomethyl)-4-(tert-butyl)cyclohexane
Formel: C₁₁H₂₁Br
Molecular Weight: 233,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847599
CAS-Nummer: 92368-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR