2-Chlorbenzimidazol
Lieferant: Apollo Scientific
Synonyme:
2-Chlor-1H-benzimidazol
OR470769-5G
APOSOR470769-5GEA
601
EUR
APOSOR470769-5G
2-Chlorbenzimidazol
2-Chlorobenzimidazole
Formel:
C₇H₅ClN₂ Molecular Weight: 152.58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051944 CAS-Nummer: 4857-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...