4-Amino-2-methylbenzyl Alcohol

Lieferant: Apollo Scientific

Synonyme: (4-Amino-2-methylphenyl)methanol

OR470770-1G
APOSOR470770-1GEA 412 EUR
APOSOR470770-1G
4-Amino-2-methylbenzyl Alcohol
4-Amino-2-methylbenzyl Alcohol
Formel: C₈H₁₁NO
Molecular Weight: 137,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110504
CAS-Nummer: 63405-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR