Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate

Lieferant: Apollo Scientific

Synonyme: (E)-Ethyl 3-(4-bromophenyl)-2-cyanoacrylate

OR470771-5G
APOSOR470771-5GEA 601 EUR
APOSOR470771-5G
Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate
Ethyl 3-(4-Bromophenyl)-2-cyanoacrylate
Formel: C₁₂H₁₀BrNO₂
Molecular Weight: 280,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00176211
CAS-Nummer: 18861-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR