1'-Boc-spiro[indane-1,4'-piperidine]

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2,3-dihydrospiro[indene-1,4'-piperidine]-1'-carboxylate

OR470772-1G
APOSOR470772-1GEA 916 EUR
APOSOR470772-1G
1'-Boc-spiro[indane-1,4'-piperidine]
1'-Boc-spiro[indane-1,4'-piperidine]
Formel: C₁₈H₂₅NO₂
Molecular Weight: 287,397 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10700122
CAS-Nummer: 148835-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR